Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 9 | 2136 | ||
NM_001297430.1 | NP_001284359.1 | 8 | 1932 | |||
NM_001297431.1 | NP_001284360.1 | 7 | 1932 | |||
NM_001297432.1 | NP_001284361.1 | 9 | 2274 | |||
NM_001297433.1 | NP_001284362.1 | 9 | 2210 | |||
NM_005882.4 | NP_005873.2 | 8 | 2013 | |||
NR_123716.1 | 7 | 1936 | ||||
XM_017007636.1 | XP_016863125.1 | 10 | 2256 | |||
XM_017007637.1 | XP_016863126.1 | 9 | 2173 | |||
GenBank mRNA | AB208907.1 | - | 3 | 5292 | ||
AF084928.1 | - | 10 | 1931 | |||
AK001088.1 | - | 9 | 2096 | |||
AK022515.1 | - | 7 | 1901 | |||
AK022586.1 | - | 8 | 1973 | |||
AK091951.1 | - | 4 | 2007 | |||
AK094253.1 | - | 6 | 1757 | |||
AK125501.1 | - | 1 | 2388 | |||
AK127875.1 | - | 5 | 2364 | |||
AK128302.1 | - | 3 | 3040 | |||
BC001225.1 | - | 9 | 2103 | 3533990 | ||
BC006470.1 | - | 9 | 2059 |