Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC93A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143947.1 | NP_001137419.1 | 7 | 1404 | ||
NM_018974.3 | NP_061847.2 | 8 | 1530 | |||
XM_011535905.2 | XP_011534207.1 | 9 | 1936 | |||
XM_011535906.2 | XP_011534208.1 | 10 | 2085 | |||
XM_011535907.2 | XP_011534209.1 | 9 | 2160 | |||
XM_017010958.1 | XP_016866447.1 | 9 | 1501 | |||
GenBank mRNA | AJ422141.1 | - | 8 | 1458 | ||
AJ508812.1 | - | 8 | 1432 | |||
AK091987.1 | - | 8 | 1530 | |||
BC098248.1 | - | 7 | 1255 | 40006812 | ||
BC099718.1 | - | 7 | 1255 | 40006815 | ||
BC105635.1 | - | 7 | 1255 | 40006813 |