Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271581.1 | NP_001258510.1 | 23 | 3080 | ||
NM_018981.2 | NP_061854.1 | 24 | 3218 | |||
NR_073365.1 | 24 | 3972 | ||||
NR_073366.1 | 25 | 4075 | ||||
NR_073367.1 | 23 | 3111 | ||||
GenBank mRNA | AF038503.1 | - | 24 | 2928 | ||
AK027450.1 | - | 22 | 2559 | |||
AK027647.1 | - | 24 | 2987 | |||
AK027701.2 | - | 21 | 2340 | |||
AK074905.1 | - | 11 | 1334 | |||
AK298138.1 | - | 19 | 2065 | |||
AL137648.1 | - | 13 | 1768 | |||
AL832267.1 | - | 17 | 1918 | |||
AL832632.2 | - | 24 | 3743 | |||
AL832646.3 | - | 22 | 2569 | |||
AY358577.1 | - | 23 | 2837 | |||
BC034713.1 | - | 4 | 511 | |||
BC070034.1 | - | 25 | 3815 | |||
BC117299.1 | - | 24 | 2933 | 40125850 | ||
BC126168.1 | - | 24 | 2933 | 8991884 |