Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 11, 12 | 1559 | ||
NM_018986.4 | NP_061859.3 | 11, 12 | 1476 | |||
NR_134639.1 | 9, 10 | 1190 | ||||
XM_006713889.2 | XP_006713952.1 | 8, 9 | 894 | |||
XM_006713891.3 | XP_006713954.1 | 5, 6 | 536 | |||
XM_011513488.1 | XP_011511790.1 | 11, 12 | 1453 | |||
XM_011513489.2 | XP_011511791.1 | 9, 10 | 1076 | |||
XM_017008300.1 | XP_016863789.1 | 11, 12 | 1585 | |||
XM_017008301.1 | XP_016863790.1 | 11, 12 | 1268 | |||
XM_017008302.1 | XP_016863791.1 | 8, 9 | 978 | |||
GenBank mRNA | AK000363.1 | - | 7, 8 | 864 | ||
AK074093.1 | - | 10, 11 | 1315 | |||
AK074402.1 | - | 11, 12 | 1353 | |||
AK126165.1 | - | 11, 12 | 1432 | |||
AK222865.1 | - | 7, 8 | 864 |