Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006144.3 | NP_006135.1 | 3 | 382 | ||
GenBank mRNA | BC015739.1 | - | 3 | 353 | 4849202 | |
BP330335.1 | - | 3 | 382 | |||
CR456968.1 | - | 3 | 345 | |||
M18737.1 | - | 3 | 345 |