Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319303.1 | NP_001306232.1 | 6 | 2362 | ||
NR_135067.1 | 5 | 1745 | ||||
NR_135068.1 | 2 | 694 | ||||
XM_017016714.1 | XP_016872203.1 | 5 | 1386 | |||
XM_017016715.1 | XP_016872204.1 | 5 | 1363 | |||
GenBank mRNA | AK172864.1 | - | 6 | 2361 | ||
BC012394.1 | - | 2 | 594 |