Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000842.4 | NP_000833.1 | 2 | 223 | ||
NM_001143831.2 | NP_001137303.1 | 1 | 25 | |||
XM_006718828.3 | XP_006718891.1 | 2 | 221 | |||
XM_011542792.1 | XP_011541094.1 | 2 | 223 | |||
XM_017017627.1 | XP_016873116.1 | 2 | 221 | |||
GenBank mRNA | AK291362.1 | - | 2 | 193 | ||
BX282658.1 | - | 2 | 195 | |||
DA316590.1 | - | 2 | 224 | |||
DR002861.1 | - | 2 | 223 |
Gene Symbol: LOC440040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027044.1 | 2 | 213 | |||
GenBank mRNA | AK294678.1 | - | 2 | 213 | ||
BC141819.1 | - | 2 | 121 | |||
BC142657.1 | - | 2 | 121 |
Set Membership: |
G-protein coupled receptor |