Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM105A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019018.2 | NP_061891.1 | 1, 2 | 185 | ||
XM_011514056.2 | XP_011512358.1 | 1, 2 | 167 | |||
GenBank mRNA | AK001989.1 | - | 1, 2 | 144 | ||
AK222731.1 | - | 1, 2 | 172 | |||
BC011524.1 | - | 1, 2 | 111 | 3872837 | ||
BX342178.2 | - | 1, 2 | 192 |