Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 3 | 637 | ||
GenBank mRNA | AJ621557.1 | - | 1 | 448 | ||
AY553630.1 | - | 3 | 652 |