Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBRS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105079.2 | NP_001098549.2 | 5 | 734 | ||
XM_011545916.2 | XP_011544218.1 | 5 | 2111 | |||
XM_011545917.2 | XP_011544219.1 | 4 | 2081 | |||
XM_011545919.2 | XP_011544221.1 | 3 | 2044 | |||
GenBank mRNA | AK096322.1 | - | 3 | 265 | ||
DB082271.1 | - | 5 | 331 | |||
DB093195.1 | - | 3 | 521 |