Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013848.3 | NP_001013870.1 | 6, 7 | 591 | ||
NM_001319194.1 | NP_001306123.1 | 6, 7 | 657 | |||
NM_001319195.1 | NP_001306124.1 | 5, 6 | 545 | |||
NM_001319200.1 | NP_001306129.1 | 5, 6 | 545 | |||
NM_019053.5 | NP_061926.3 | 5, 6 | 545 | |||
XM_017016344.1 | XP_016871833.1 | 6, 7 | 652 | |||
XM_017016345.1 | XP_016871834.1 | 5, 6 | 626 | |||
XM_017016346.1 | XP_016871835.1 | 5, 6 | 626 | |||
XM_017016347.1 | XP_016871836.1 | 5, 6 | 829 | |||
XM_017016348.1 | XP_016871837.1 | 6, 7 | 652 | |||
GenBank mRNA | AK128190.1 | - | 6, 7 | 591 | ||
AK293857.1 | - | 5, 6 | 508 | |||
AK300285.1 | - | 6, 7 | 657 | |||
AK315544.1 | - | 5, 6 | 480 | |||
AL832501.1 | - | 5, 6 | 489 | |||
BC028395.2 | - | 5, 6 | 480 | 4820245 | ||
DA187840.1 | - | 5, 6 | 532 | |||
DA333847.1 | - | 5, 6 | 546 | |||
EF571007.1 | - | 5, 6 | 486 |