Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105663.2 | NP_001099133.1 | 4 | 892 | ||
NM_001243657.1 | NP_001230586.1 | 5 | 986 | |||
NM_001243660.1 | NP_001230589.1 | 4 | 847 | |||
NM_001243661.1 | NP_001230590.1 | 3 | 733 | |||
XM_011523032.2 | XP_011521334.1 | 5 | 1010 | |||
XM_011523033.2 | XP_011521335.1 | 6 | 1050 | |||
XM_017023170.1 | XP_016878659.1 | 6 | 1104 | |||
GenBank mRNA | AK296963.1 | - | 3 | 706 |