Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135998.2 | NP_001129470.1 | 1 | 725 | ||
NM_019056.6 | NP_061929.2 | 1 | 725 | |||
GenBank mRNA | AF044213.1 | - | 1 | 212 | ||
AF134476.1 | - | 1 | 259 | |||
AF251063.1 | - | 1 | 558 | |||
AK000501.1 | - | 1 | 207 | |||
AY359056.1 | - | 1 | 725 | |||
BC010665.1 | - | 1 | 533 | 3870660 | ||
BC107805.1 | - | 1 | 208 | 6496406 | ||
CR457239.1 | - | 1 | 194 |