Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024019.3 | 8, 9 | 3069 | |||
XM_006714875.2 | XP_006714938.1 | 6, 7 | 1538 | |||
XM_006714876.1 | XP_006714939.1 | 6, 7 | 2261 | |||
XM_006714878.1 | XP_006714941.1 | 7, 8 | 2115 | |||
XM_006714879.3 | XP_006714942.1 | 7, 8 | 1358 | |||
XM_006714880.1 | XP_006714943.1 | 6, 7 | 2156 | |||
XM_006714884.1 | XP_006714947.1 | 4, 5 | 659 | |||
XM_006714885.1 | XP_006714948.1 | 3, 4 | 463 | |||
XM_011534572.1 | XP_011532874.1 | 6, 7 | 1433 | |||
XM_011534573.1 | XP_011532875.1 | 7, 8 | 1533 | |||
XM_011534574.1 | XP_011532876.1 | 7, 8 | 1978 | |||
XM_011534579.2 | XP_011532881.1 | 6, 7 | 1297 | |||
XM_011534580.1 | XP_011532882.1 | 7, 8 | 2322 | |||
XM_011534581.1 | XP_011532883.1 | 6, 7 | 2002 | |||
XM_011534582.1 | XP_011532884.1 | 7, 8 | 2509 | |||
XM_011534583.1 | XP_011532885.1 | 7, 8 | 2250 | |||
GenBank mRNA | AB067518.1 | - | 8, 9 | 3026 | ||
AK001266.1 | - | 2, 3 | 339 | |||
AK096032.1 | - | 6, 7 | 1221 | |||
BC008784.2 | - | 4, 5 | 659 | 3626782 | ||
BC086859.1 | - | 4, 5 | 883 | |||
BX647587.1 | - | 5, 6 | 1154 |