Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 2 | 408 | ||
NM_019063.4 | NP_061936.2 | 2 | 408 | |||
XM_005264267.1 | XP_005264324.1 | 2 | 388 | |||
XM_005264268.3 | XP_005264325.1 | 2 | 388 | |||
XM_006711991.1 | XP_006712054.1 | 2 | 388 | |||
XM_006711992.2 | XP_006712055.1 | 3 | 509 | |||
GenBank mRNA | AF177377.2 | - | 2 | 370 | ||
AK001804.1 | - | 1451 | ||||
AK314702.1 | - | 2 | 382 | |||
BC104647.1 | - | 2 | 335 | |||
BC140845.1 | - | 2 | 331 | 9021713 | ||
BC146799.1 | - | 2 | 382 | 8860304 | ||
DB471322.1 | - | 2 | 398 | |||
HY044849.1 | - | 2 | 409 |