Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195831.2 | NP_001182760.2 | 16 | 2781 | ||
XM_005270520.1 | XP_005270577.1 | 16 | 2758 | |||
XM_006710383.1 | XP_006710446.1 | 17 | 2847 | |||
XM_011540793.1 | XP_011539095.1 | 16 | 2781 | |||
XM_011540794.1 | XP_011539096.1 | 16 | 2713 | |||
XM_011540795.2 | XP_011539097.1 | 16 | 2705 | |||
XM_011540796.1 | XP_011539098.1 | 17 | 2788 | |||
XM_011540797.2 | XP_011539099.1 | 16 | 3062 | |||
XM_011540798.1 | XP_011539100.1 | 15 | 2545 | |||
XM_011540799.1 | XP_011539101.1 | 14 | 2371 | |||
XM_011540800.1 | XP_011539102.1 | 16 | 2758 | |||
XM_017000421.1 | XP_016855910.1 | 15 | 2584 | |||
XM_017000422.1 | XP_016855911.1 | 16 | 2758 | |||
GenBank mRNA | DV080053.1 | - | 2 | 278 |