Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019087.2 | NP_061960.1 | 2 | 226 | ||
XM_011543498.2 | XP_011541800.1 | 4 | 331 | |||
XM_011543499.2 | XP_011541801.1 | 3 | 274 | |||
XM_011543500.2 | XP_011541802.1 | 3 | 300 | |||
XM_017009598.1 | XP_016865087.1 | 4 | 318 | |||
GenBank mRNA | AK000058.1 | - | 2 | 226 | ||
AK290886.1 | - | 2 | 227 | |||
BC026093.1 | - | 2 | 230 | 4699068 | ||
CR457225.1 | - | 2 | 132 |