Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 21 | 4417 | ||
NM_001277444.1 | NP_001264373.1 | 25 | 4924 | |||
XM_006711317.2 | XP_006711380.1 | 26 | 5152 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF419616.1 | - | 23 | 4315 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 22 | 4663 | ||
NM_183372.5 | NP_899228.4 | 24 | 5357 | |||
NR_046188.3 | 25 | 5425 | ||||
NR_110864.1 | 23 | 5349 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 66 | 9970 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 22 | 4688 | ||
NM_173638.4 | NP_775909.2 | 19 | 4158 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 18340 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 20 | 3718 | |||
GenBank mRNA | AK125792.1 | - | 20 | 3718 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 20 | 4514 | ||
NR_102404.1 | 19 | 4350 | ||||
NR_102405.1 | 18 | 4247 |