Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125967.1 | 4 | 919 | |||
NR_125968.1 | 3 | 624 |
Gene Symbol: LOC100996724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003064.1 | XP_016858553.1 | 46 | 7279 |
Gene Symbol: LOC101060254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003057.1 | XP_016858546.1 | 14 | 1948 |