Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271870.2 | NP_001258799.2 | 1 | 382 | ||
XM_005277421.3 | XP_005277478.1 | 1 | 461 | |||
XM_017002085.1 | XP_016857574.1 | 1 | 461 | |||
XM_017002087.1 | XP_016857576.1 | 1 | 461 | |||
XM_017002088.1 | XP_016857577.1 | 1 | 461 | |||
XM_017002089.1 | XP_016857578.1 | 1 | 461 | |||
XM_017002090.1 | XP_016857579.1 | 1 | 461 | |||
XM_017002091.1 | XP_016857580.1 | 1 | 461 | |||
XM_017002092.1 | XP_016857581.1 | 1 | 461 | |||
GenBank mRNA | DA326673.1 | - | 1 | 382 |
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 1 | 463 | ||
NM_001300952.1 | NP_001287881.1 | 1 | 463 | |||
NM_015326.4 | NP_056141.2 | 1 | 463 | |||
XM_005277510.2 | XP_005277567.1 | 2 | 650 | |||
XM_005277511.2 | XP_005277568.1 | 2 | 650 | |||
XM_005277514.1 | XP_005277571.1 | 2 | 650 | |||
XM_005277515.1 | XP_005277572.1 | 2 | 650 | |||
XM_011509354.1 | XP_011507656.1 | 2 | 650 | |||
XM_011509355.1 | XP_011507657.1 | 2 | 650 | |||
XM_011509356.2 | XP_011507658.1 | 2 | 650 | |||
XM_011509359.1 | XP_011507661.1 | 2 | 650 | |||
XM_017000838.1 | XP_016856327.1 | 2 | 650 | |||
XM_017000841.1 | XP_016856330.1 | 2 | 650 | |||
GenBank mRNA | AK124556.1 | - | 1 | 2004 | ||
AK295845.1 | - | 1 | 219 | |||
BC150646.1 | - | 1 | 416 | |||
DA110905.1 | - | 2 | 551 |
Gene Symbol: SRGAP2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271872.1 | NP_001258801.1 | 2 | 526 | ||
XM_005277497.3 | XP_005277554.1 | 2 | 621 | |||
XM_017002149.1 | XP_016857638.1 | 2 | 621 | |||
GenBank mRNA | BC112927.1 | - | 2 | 526 |
Gene Symbol: SRGAP2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120535.1 | 2 | 592 |