Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316764.1 | NP_001303693.1 | 3 | 531 | ||
NM_001316765.1 | NP_001303694.1 | 3 | 384 | |||
NM_001316766.1 | NP_001303695.1 | 2 | 614 | |||
GenBank mRNA | BU184705.1 | - | 2 | 575 | ||
HY047622.1 | - | 3 | 385 |