Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012089.2 | NP_036221.2 | 13 | 2269 | ||
XM_011544135.2 | XP_011542437.1 | 13 | 3040 | |||
XM_011544136.2 | XP_011542438.1 | 11 | 1506 | |||
GenBank mRNA | AB013380.1 | - | 13 | 2256 | ||
AF216833.1 | - | 13 | 2269 | |||
AK172794.1 | - | 12 | 1525 | |||
BC064930.1 | - | 13 | 2264 | 6143235 | ||
DQ786241.1 | - | 6 | 647 | |||
U18237.1 | - | 6 | 699 |