Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114357.1 | NP_001107829.1 | 2 | 232 | ||
XM_005263019.3 | XP_005263076.1 | 4 | 585 | |||
XM_011531989.2 | XP_011530291.1 | 3 | 511 | |||
XM_017008236.1 | XP_016863725.1 | 4 | 1978 | |||
XM_017008237.1 | XP_016863726.1 | 5 | 753 | |||
XM_017008238.1 | XP_016863727.1 | 3 | 1762 | |||
XM_017008239.1 | XP_016863728.1 | 3 | 1765 | |||
XM_017008240.1 | XP_016863729.1 | 3 | 514 | |||
XM_017008242.1 | XP_016863731.1 | 4 | 2044 | |||
GenBank mRNA | AY947525.1 | - | 1 | 102 | ||
BC150574.1 | - | 2 | 232 | 40127870 |