Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021490.2 | 2 | 213 | |||
NR_021491.2 | 2 | 213 | ||||
NR_109818.1 | 3 | 326 | ||||
NR_109819.1 | 2 | 888 | ||||
NR_109820.1 | 2 | 213 | ||||
NR_109821.1 | 2 | 213 | ||||
NR_109822.1 | 2 | 888 | ||||
NR_109823.1 | 2 | 210 | ||||
NR_109824.1 | 2 | 213 | ||||
NR_109825.1 | 3 | 993 | ||||
GenBank mRNA | AK095719.1 | - | 3 | 304 | ||
AK124699.1 | - | 2 | 938 | |||
AK296281.1 | - | 2 | 575 | |||
AK311143.1 | - | 2 | 112 | |||
AK311323.1 | - | 2 | 601 | |||
BC010610.1 | - | 2 | 181 | |||
BC018089.1 | - | 2 | 190 | |||
BC042879.1 | - | 2 | 112 | |||
BC044619.1 | - | 3 | 469 | |||
BC048999.1 | - | 3 | 469 | |||
BC144330.1 | - | 2 | 1488 | |||
BC144331.1 | - | 2 | 1488 | |||
DA718791.1 | - | 2 | 196 | |||
DB257894.1 | - | 2 | 112 |