Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021490.2 | 4 | 461 | |||
NR_021491.2 | 4 | 461 | ||||
NR_109818.1 | 5 | 574 | ||||
NR_109819.1 | 4 | 1136 | ||||
NR_109820.1 | 4 | 461 | ||||
NR_109821.1 | 4 | 461 | ||||
NR_109823.1 | 4 | 458 | ||||
NR_109824.1 | 4 | 461 | ||||
NR_109825.1 | 5 | 1241 | ||||
GenBank mRNA | AK095719.1 | - | 5 | 552 | ||
AK124699.1 | - | 4 | 1186 | |||
AK311323.1 | - | 4 | 849 | |||
BC010610.1 | - | 4 | 429 | |||
BC018089.1 | - | 4 | 438 | |||
BC042879.1 | - | 4 | 360 | |||
BC044619.1 | - | 5 | 717 | |||
BC048999.1 | - | 5 | 717 | |||
BC144330.1 | - | 4 | 1736 | |||
BC144331.1 | - | 4 | 1736 | |||
BX647510.1 | - | 3 | 229 | |||
DA718791.1 | - | 4 | 444 | |||
DB257894.1 | - | 4 | 360 | |||
HY108556.1 | - | 4 | 391 |