Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021490.2 | 1 | 148 | |||
NR_021491.2 | 1 | 148 | ||||
NR_109818.1 | 1 | 148 | ||||
NR_109819.1 | 1 | 148 | ||||
NR_109820.1 | 1 | 148 | ||||
NR_109821.1 | 1 | 148 | ||||
NR_109822.1 | 1 | 148 | ||||
NR_109823.1 | 1 | 148 | ||||
NR_109824.1 | 1 | 148 | ||||
GenBank mRNA | AK095719.1 | - | 1 | 126 | ||
AK311143.1 | - | 1 | 47 | |||
BC010610.1 | - | 1 | 116 | |||
BC018089.1 | - | 1 | 125 | |||
BC042879.1 | - | 1 | 47 | |||
DA107272.1 | - | 1 | 387 | |||
DA718791.1 | - | 1 | 131 | |||
DB257894.1 | - | 1 | 47 | |||
DC298450.1 | - | 1 | 294 | |||
HY108556.1 | - | 1 | 81 |