Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001483.2 | NP_001001483.1 | 2 | 181 | ||
NM_019028.2 | NP_061901.2 | 3 | 327 | |||
XM_005252996.2 | XP_005253053.1 | 3 | 299 | |||
XM_011520193.1 | XP_011518495.1 | 1 | 24 | |||
XM_011520194.2 | XP_011518496.1 | 3 | 302 | |||
XM_011520195.1 | XP_011518497.1 | 3 | 237 | |||
GenBank mRNA | AB024495.1 | - | 3 | 275 | ||
AB097014.1 | - | 3 | 325 | |||
AB097027.1 | - | 3 | 325 | |||
AK001714.1 | - | 3 | 325 | |||
BC036020.1 | - | 3 | 312 | 5272445 | ||
BC050690.1 | - | 2 | 181 | 6171322 | ||
BC056152.1 | - | 3 | 327 | 6170701 | ||
BX648156.1 | - | 2 | 171 |