Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100132304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120493.1 | 2 | 176 | |||
NR_120494.1 | 2 | 164 | ||||
GenBank mRNA | AA436071.1 | - | 2 | 113 | ||
HY042459.1 | - | 2 | 122 |
Gene Symbol: LINC00684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034116.1 | 2 | 176 | |||
NR_120499.1 | 2 | 164 | ||||
GenBank mRNA | AA393776.1 | - | 2 | 132 |