Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145076.2 | NP_001138548.1 | 3 | 589 | ||
NM_019063.4 | NP_061936.2 | 3 | 589 | |||
XM_005264267.1 | XP_005264324.1 | 3 | 569 | |||
XM_005264268.3 | XP_005264325.1 | 3 | 569 | |||
XM_006711991.1 | XP_006712054.1 | 3 | 569 | |||
XM_006711992.2 | XP_006712055.1 | 4 | 690 | |||
GenBank mRNA | AF177377.2 | - | 3 | 551 | ||
AK001804.1 | - | 1632 | ||||
AK314702.1 | - | 3 | 563 | |||
BC104647.1 | - | 3 | 516 | |||
BC140845.1 | - | 3 | 512 | 9021713 | ||
BC146799.1 | - | 3 | 563 | 8860304 |