Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291712.1 | NP_001278641.1 | 1 | 361 | ||
NM_001291723.1 | NP_001278652.1 | 1 | 361 | |||
NR_073496.1 | 1 | 361 | ||||
XM_017021079.1 | XP_016876568.1 | 1 | 442 | |||
XM_017021080.1 | XP_016876569.1 | 1 | 442 | |||
XM_017021081.1 | XP_016876570.1 | 1 | 442 | |||
XM_017021084.1 | XP_016876573.1 | 1 | 442 | |||
GenBank mRNA | BX247957.1 | - | 1 | 410 | ||
CN366005.1 | - | 1 | 361 |