Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019071.2 | NP_061944.2 | 3 | 315 | ||
NM_198267.1 | NP_938008.1 | 3 | 315 | |||
XM_017012369.1 | XP_016867858.1 | 3 | 323 | |||
GenBank mRNA | AF074968.1 | - | 3 | 292 | ||
AF161419.1 | - | 3 | 305 | |||
AF180298.1 | - | 3 | 282 | |||
AK000096.1 | - | 3 | 316 | |||
AK291905.1 | - | 3 | 293 | |||
AK293682.1 | - | 2 | 201 | |||
AL603623.1 | - | 3 | 292 | |||
AY007790.1 | - | 3 | 296 | |||
BC009776.1 | - | 3 | 270 | |||
BC009777.1 | - | 3 | 303 | |||
BC010851.1 | - | 3 | 262 | |||
BC062634.1 | - | 3 | 278 | |||
BC073865.1 | - | 3 | 283 | |||
BC093091.1 | - | 3 | 299 | |||
BC093689.1 | - | 3 | 275 | |||
BC101609.1 | - | 3 | 275 | |||
BC143316.1 | - | 3 | 275 |