Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019074.3 | NP_061947.1 | 9 | 2038 | ||
GenBank mRNA | AB036931.1 | - | 9 | 1978 | ||
AB043894.1 | - | 9 | 1702 | |||
AF253468.1 | - | 9 | 1702 | |||
AF279305.1 | - | 8 | 1608 | |||
AK313831.1 | - | 9 | 2033 | |||
AY358894.1 | - | 9 | 1737 | |||
BC106950.2 | - | 9 | 1838 | 40034887 |