Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256826.1 | NP_001243755.1 | 6 | 804 | ||
NM_019088.3 | NP_061961.2 | 7 | 834 | |||
NR_046384.1 | 6 | 712 | ||||
GenBank mRNA | AJ401156.1 | - | 7 | 659 | ||
AK001985.1 | - | 7 | 731 | |||
AK098423.1 | - | 6 | 601 | |||
AK294625.1 | - | 6 | 634 | |||
BC000017.2 | - | 7 | 678 | 3504227 | ||
BC013402.1 | - | 7 | 750 | 2960015 |