Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000839.1 | XP_016856328.1 | 1 | 645 |
Gene Symbol: SRGAP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002089.1 | XP_016857578.1 | 4 | 891 | ||
XM_017002091.1 | XP_016857580.1 | 4 | 891 |