Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286216.1 | NP_001273145.1 | 4 | 386 | ||
NM_019096.4 | NP_061969.3 | 4 | 581 | |||
XM_017010976.1 | XP_016866465.1 | 4 | 575 | |||
GenBank mRNA | AB024574.1 | - | 5 | 424 | ||
AF168990.1 | - | 4 | 378 | |||
AK290267.1 | - | 4 | 386 | |||
AL834331.1 | - | 4 | 363 | |||
BC028347.2 | - | 2 | 289 | |||
BC064968.1 | - | 4 | 534 | 6149902 |