Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003692.1 | NP_001003692.1 | 2 | 249 | ||
NM_001318129.1 | NP_001305058.1 | 2 | 203 | |||
NM_019103.2 | NP_061976.1 | 3 | 364 | |||
GenBank mRNA | AA312132.1 | - | 2 | 249 | ||
AK123261.1 | - | 2 | 203 | |||
AK292671.1 | - | 2 | 230 | |||
AL160311.1 | - | 2 | 248 | |||
BC009717.1 | - | 2 | 214 | 3890730 | ||
BG285784.1 | - | 3 | 328 | |||
BQ931527.1 | - | 2 | 177 | |||
BU598307.1 | - | 2 | 251 | |||
CR456353.1 | - | 1 | 136 | |||
CR457393.1 | - | 1 | 113 | |||
DT217179.1 | - | 3 | 380 |