Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306188.1 | NP_001293117.1 | 4 | 590 | ||
NM_019556.2 | NP_062456.1 | 4 | 590 | |||
XM_011531366.2 | XP_011529668.1 | 4 | 544 | |||
XM_017029653.1 | XP_016885142.1 | 5 | 783 | |||
XM_017029654.1 | XP_016885143.1 | 4 | 544 | |||
GenBank mRNA | AK299074.1 | - | 4 | 503 | ||
AK307196.1 | - | 3 | 417 | |||
AK316570.1 | - | 4 | 465 | |||
AL137163.1 | - | 4 | 513 | |||
BC005700.1 | - | 4 | 493 | 3450155 | ||
CB997365.1 | - | 4 | 498 | |||
CN428237.1 | - | 4 | 592 | |||
DQ323992.1 | - | 3 | 326 |