Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 3 | 587 | |||
NR_110066.1 | 2 | 269 | ||||
NR_110067.1 | 3 | 525 | ||||
NR_110070.1 | 4 | 376 | ||||
GenBank mRNA | AK090593.1 | - | 2 | 151 | ||
BG478902.1 | - | 270 | ||||
BM544796.1 | - | 4 | 397 | |||
BM722001.1 | - | 3 | 506 | |||
DA025114.1 | - | 2 | 151 | |||
HY110280.1 | - | 3 | 457 |