Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185149.1 | NP_001172078.1 | 1 | 241 |
Gene Symbol: CLDN22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111319.1 | NP_001104789.1 | 1 | 797 | ||
GenBank mRNA | AK098064.1 | - | 1 | 191 | ||
AK303864.1 | - | 291 |