Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YLPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019589.2 | NP_062535.2 | 2 | 1055 | ||
XM_005267860.3 | XP_005267917.1 | 2 | 1099 | |||
XM_011536966.2 | XP_011535268.1 | 3 | 214 | |||
XM_011536967.1 | XP_011535269.1 | 2 | 1099 | |||
GenBank mRNA | AK095760.1 | - | 2 | 1055 | ||
BC044636.1 | - | 2 | 416 | |||
DB096717.1 | - | 1 | 41 |