Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080425.3 | NP_001073894.1 | 3 | 830 | ||
NM_001127688.2 | NP_001121160.1 | 2 | 747 | |||
GenBank mRNA | AK000959.1 | - | 3 | 785 | ||
AL544124.3 | - | 2 | 723 | |||
AY833563.1 | - | 3 | 785 |