Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 6 | 592 | ||
XM_005260768.2 | XP_005260825.1 | 6 | 607 | |||
XM_006723598.1 | XP_006723661.1 | 6 | 607 | |||
XM_006723599.1 | XP_006723662.1 | 6 | 607 | |||
XM_011529309.1 | XP_011527611.1 | 6 | 607 | |||
XM_011529310.2 | XP_011527612.1 | 4 | 228 | |||
XM_017027972.1 | XP_016883461.1 | 6 | 607 | |||
XM_017027973.1 | XP_016883462.1 | 4 | 209 | |||
XM_017027974.1 | XP_016883463.1 | 4 | 204 | |||
XM_017027975.1 | XP_016883464.1 | 6 | 511 | |||
XM_017027976.1 | XP_016883465.1 | 4 | 364 | |||
XM_017027977.1 | XP_016883466.1 | 6 | 607 | |||
GenBank mRNA | BC171842.1 | - | 4 | 364 | ||
CR990457.1 | - | 4 | 428 | |||
DQ310704.1 | - | 6 | 449 |