Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020722.1 | NP_065773.1 | 9, 10 | 3762 | ||
XM_005265752.2 | XP_005265809.1 | 8, 9 | 3769 | |||
XM_005265755.2 | XP_005265812.1 | 5, 6 | 3198 | |||
XM_005265756.2 | XP_005265813.1 | 5, 6 | 3246 | |||
XM_006714045.2 | XP_006714108.1 | 5, 6 | 3223 | |||
XM_011534397.2 | XP_011532699.1 | 8, 9 | 4512 | |||
XM_017008473.1 | XP_016863962.1 | 8, 9 | 3498 | |||
XM_017008474.1 | XP_016863963.1 | 7, 8 | 4376 | |||
GenBank mRNA | AB033037.1 | - | 3, 4 | 2083 | ||
AK097530.1 | - | 6, 7 | 2292 | |||
AK126014.1 | - | 9, 10 | 3762 | |||
AK302486.1 | - | 6, 7 | 3200 | |||
AL133028.1 | - | 2, 3 | 2174 | |||
AL137336.1 | - | 2, 3 | 1540 | |||
BC144536.1 | - | 8, 9 | 3516 | 9053063 | ||
BC150615.1 | - | 8, 9 | 3516 | 9056985 | ||
BC171783.1 | - | 8, 9 | 3498 | 9054437 |