Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020846.1 | NP_065897.1 | 2 | 555 | ||
XM_005248121.2 | XP_005248178.1 | 2 | 671 | |||
GenBank mRNA | AB040891.1 | - | 2 | 525 | ||
AK026476.1 | - | 2 | 102 | |||
AK291006.1 | - | 2 | 685 | |||
BC040993.1 | - | 2 | 834 | 5272534 |