Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003367.3 | 3 | 770 | |||
GenBank mRNA | BC041065.1 | - | 2 | 529 | ||
M34428.1 | - | 3 | 745 |