Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021241.2 | NP_067064.2 | 4 | 1176 | ||
XM_005260004.4 | XP_005260061.1 | 9 | 4411 | |||
XM_005260005.4 | XP_005260062.1 | 8 | 4240 | |||
XM_005260006.4 | XP_005260063.1 | 8 | 4012 | |||
XM_005260007.4 | XP_005260064.1 | 7 | 3841 | |||
XM_005260008.2 | XP_005260065.1 | 7 | 2255 | |||
XM_005260009.2 | XP_005260066.1 | 5 | 1602 | |||
XM_005260010.3 | XP_005260067.1 | 5 | 1615 | |||
XM_005260011.2 | XP_005260068.1 | 5 | 1685 | |||
XM_005260012.2 | XP_005260069.1 | 4 | 1203 | |||
XM_006722828.3 | XP_006722891.1 | 7 | 2024 | |||
XM_011528163.2 | XP_011526465.1 | 5 | 1676 | |||
GenBank mRNA | AK027615.1 | - | 1 | 130 | ||
AK091183.1 | - | 2 | 636 | |||
AK122890.1 | - | 5 | 1232 | |||
AK131404.1 | - | 4 | 1177 | |||
BC098445.1 | - | 1 | 410 | |||
BC136641.1 | - | 3 | 1109 | 9020631 | ||
BC144332.1 | - | 4 | 1508 | 9052852 |