Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021241.2 | NP_067064.2 | 8 | 2640 | ||
XM_005260004.4 | XP_005260061.1 | 13 | 5875 | |||
XM_005260005.4 | XP_005260062.1 | 12 | 5704 | |||
XM_005260006.4 | XP_005260063.1 | 12 | 5476 | |||
XM_005260007.4 | XP_005260064.1 | 11 | 5305 | |||
XM_005260008.2 | XP_005260065.1 | 11 | 3719 | |||
XM_005260009.2 | XP_005260066.1 | 9 | 3066 | |||
XM_005260010.3 | XP_005260067.1 | 9 | 3079 | |||
XM_005260011.2 | XP_005260068.1 | 9 | 3149 | |||
XM_005260012.2 | XP_005260069.1 | 8 | 2667 | |||
XM_006722828.3 | XP_006722891.1 | 11 | 3488 | |||
XM_011528163.2 | XP_011526465.1 | 9 | 3140 | |||
GenBank mRNA | AK027615.1 | - | 5 | 1594 | ||
AK091183.1 | - | 6 | 2100 | |||
AK122890.1 | - | 9 | 2696 | |||
AK131404.1 | - | 8 | 2641 | |||
AL390184.1 | - | 3 | 512 | |||
BC002329.2 | - | 3 | 503 | |||
BC007551.1 | - | 4 | 1152 | |||
BC062360.1 | - | 5 | 1364 | |||
BC098445.1 | - | 5 | 1874 | |||
BC136641.1 | - | 7 | 2573 | 9020631 | ||
BC144332.1 | - | 8 | 2972 | 9052852 |