Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293725.1 | NP_001280654.1 | 15 | 3431 | ||
NM_005640.2 | NP_005631.1 | 15 | 3416 | |||
NR_121653.1 | 16 | 3536 | ||||
XM_017025933.1 | XP_016881422.1 | 15 | 3040 | |||
GenBank mRNA | AY888048.1 | - | 15 | 3416 | ||
AY888049.1 | - | 16 | 3537 |