Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080432.2 | NP_001073901.1 | 6 | 1310 | ||
XM_011523313.2 | XP_011521615.1 | 6 | 1304 | |||
XM_011523314.2 | XP_011521616.1 | 6 | 1304 | |||
XM_011523315.2 | XP_011521617.1 | 6 | 1304 | |||
XM_011523316.2 | XP_011521618.1 | 6 | 1304 | |||
XM_017023654.1 | XP_016879143.1 | 6 | 1304 | |||
XM_017023655.1 | XP_016879144.1 | 6 | 1274 | |||
XM_017023656.1 | XP_016879145.1 | 6 | 1274 | |||
XM_017023657.1 | XP_016879146.1 | 6 | 1274 | |||
GenBank mRNA | AB051539.1 | - | 6 | 1094 | ||
AK096554.1 | - | 6 | 1117 | |||
AK295196.1 | - | 5 | 463 |