Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080432.2 | NP_001073901.1 | 9 | 2697 | ||
XM_011523313.2 | XP_011521615.1 | 9 | 2691 | |||
GenBank mRNA | AB051539.1 | - | 9 | 2481 | ||
AK096554.1 | - | 9 | 2504 | |||
BC003583.1 | - | 3 | 1208 | |||
BC030798.1 | - | 3 | 1465 | |||
BC137091.1 | - | 4 | 1380 |